Läs mer
4-Methylpentanoyl Chloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-Methylpentanoyl Chloride |
| CAS | 38136-29-7 |
| Molekylformel | C6H11ClO |
| Kvantitet | 500 mg |
| Synonym | 4-Methylpentanoyl Chloride, 4-Methylvaleric Acid Chloride, 4-Methylvaleroyl Chloride, 4-Methylvaleryl Chloride, Isocaproyl Chloride, Isohexanoyl Chloride, γ-Methylvaleroyl Chloride, γ-Methylvaleryl Chloride |
| InChI formel | InChI=1S/C6H11ClO/c1-5(2)3-4-6(7)8/h5H,3-4H2,1-2H3 |
| LEDER | CC(C)CCC(=O)Cl |
| Molekylvikt (g/mol) | 134.6 |