Läs mer
(4-Methylphenyl)(phenyl)acetonitrile, TRC
Produktkod. 30471640
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30471640 25 mg
1 options
Produktkod. 30471640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM01487625MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4-Methylphenyl)(phenyl)acetonitrile
CAS 6974-49-8
Molekylformel C15H13N
Kvantitet 25 mg
Synonym 2-phenyl-2-(p-tolyl)acetonitrile, 4-Methyl-α-phenylbenzeneacetonitrile (ACI), Acetonitrile, phenyl-p-tolyl- (8CI), (4-Methylphenyl)phenylacetonitrile, (p-Methylphenyl)phenylacetonitrile, 2-(4-Methylphenyl)-2-phenylacetonitrile, NSC 23824
InChI formel InChI=1S/C15H13N/c1-12-7-9-14(10-8-12)15(11-16)13-5-3-2-4-6-13/h2-10,15H,1H3
LEDER N#CC(C=1C=CC=CC1)C2=CC=C(C=C2)C
IUPAC-namn 4-Methyl-α-phenylbenzeneacetonitrile
Molekylvikt (g/mol) 207.27
Formel vikt 207.11