Läs mer
4-Methylsyringol Gentiobioside, TRC
Produktkod. 30136963
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136963 5 mg
30135002 10 mg
30005214 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136963 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM4207505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1416253-73-0
Molekylformel C21 H32 O13
Molekylvikt (g/mol) 492.47
Synonym (2S,3R,4S,5S,6R)-2-(2,6-Dimethoxy-4-methylphenoxy)-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol
IUPAC-namn (2R,3R,4S,5S,6R)-2-[[(2R,3S,4S,5R,6S)-6-(2,6-dimethoxy-4-methyl-phenoxy)-3,4,5-trihydroxy-tetrahydropyran-2-yl]methoxy]-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol
LEDER COc1cc(C)cc(OC)c1O[C@@H]2O[C@H](CO[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material 4-Methylsyringol Gentiobioside
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C21H32O13/c1-8-4-9(29-2)19(10(5-8)30-3)34-21-18(28)16(26)14(24)12(33-21)7-31-20-17(27)15(25)13(23)11(6-22)32-20/h4-5,11-18,20-28H,6-7H2,1-3H3/t11-,12-,13-,14-,15+,16+,17-,18-,20-,21+/m1/s1
Formel vikt 492.1843
Procent renhet >95%