missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Melting Point | 287.0°C |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 100 g |
Formula Weight | 143.17 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methylthiazole-5-carboxylic acid |
Safety and Handling
Product Identifier
- 4-Methylthiazole-5-carboxylic acid
- Warning
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'