Läs mer
4-(Methylthio)butylnitrile, TRC
Produktkod. 30475900
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475900 10 mg
30481124 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30475900 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33070010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59121-24-3
Molekylformel C5 H9 N S
Molekylvikt (g/mol) 115.2
IUPAC-namn 4-methylsulfanylbutanenitrile
LEDER CSCCCC#N
Specifikationer
Kemiskt namn eller material 4-(Methylthio)butylnitrile
Kvantitet 10 mg
InChI formel InChI=1S/C5H9NS/c1-7-5-3-2-4-6/h2-3,5H2,1H3
Formel vikt 115.0456