Läs mer
4-Methylumbelliferyl 2-Acetamido-2-deoxy-Alpha-D-galactopyranoside, TRC
Produktkod. 30470129
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470129 100 mg
30465872 10 mg
30465429 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30470129 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM333500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 124223-99-0
Molekylformel C18 H21 N O8
Molekylvikt (g/mol) 379.36
Synonym 2H-1-Benzopyran-2-one, 7-[[2-(acetylamino)-2-deoxy-α-D-galactopyranosyl]oxy]-4-methyl-: 7-[[2-(Acetylamino)-2-deoxy-α-D-galactopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one, 4-Methylumbelliferyl 2-acetamido-2-deoxy-α-D-galactopyranoside, 4-Methylumbelliferyl N-acetyl-α-D-galactosaminide, 4-Methylumbelliferyl N-acetyl-α-galactosaminide, 4-Methylumbelliferyl α-D-N-acetylgalactosaminide, 4-Methylumbelliferyl α-D-acetylgalactosaminide
IUPAC-namn N-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]acetamide
LEDER CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@@H]1Oc2ccc3C(=CC(=O)Oc3c2)C
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl 2-Acetamido-2-deoxy-alpha-D-galactopyranoside
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)/t13-,15-,16+,17-,18+/m1/s1
Formel vikt 379.1267
Procent renhet >95%