Läs mer
4-Methylumbelliferyl 2-Acetamido-2-deoxy-Alpha-D-glucopyranoside, TRC
Produktkod. 30005503
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005503 100 mg
30005553 50 mg
30007050 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005503 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM333800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80265-04-9
Molekylformel C18 H21 N O8
Molekylvikt (g/mol) 379.36
Synonym 2H-1-Benzopyran-2-one, 7-[[2-(acetylamino)-2-deoxy-α-D-glucopyranosyl]oxy]-4-methyl-, 7-[[2-(Acetylamino)-2-deoxy-α-D-glucopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one, 4-Methylumbelliferyl N-acetyl-α-D-glucosaminide, 4-Methylumbelliferyl N-acetyl-α-glucosaminide, 4-Methylumbelliferyl α-D-N-acetylglucosaminide, 4-Methylumbelliferyl α-D-acetylglucosaminide, 4-Methylumbelliferyl 2-acetamido-2-deoxy-α-D-glucopyranoside
IUPAC-namn N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]acetamide
LEDER CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1Oc2ccc3C(=CC(=O)Oc3c2)C
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl 2-Acetamido-2-deoxy-a-D-glucopyranoside
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)/t13-,15-,16-,17-,18+/m1/s1
Formel vikt 379.1267
Procent renhet >95%