Läs mer
4-Methylumbelliferyl 2-Amino-2-deoxy-Alpha-D-glucopyranoside (>90%), TRC
Produktkod. 30478953
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478953 5 mg
30471115 25 mg
30470872 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30478953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3342755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 137687-00-4
Molekylformel C16 H19 N O7
Molekylvikt (g/mol) 337.32
IUPAC-namn 7-[(2R,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-methylchromen-2-one
LEDER CC1=CC(=O)Oc2cc(O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3N)ccc12
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl 2-Amino-2-deoxy-a-D-glucopyranoside
Kvantitet 5 mg
InChI formel InChI=1S/C16H19NO7/c1-7-4-12(19)23-10-5-8(2-3-9(7)10)22-16-13(17)15(21)14(20)11(6-18)24-16/h2-5,11,13-16,18,20-21H,6,17H2,1H3/t11-,13-,14-,15-,16+/m1/s1
Formel vikt 337.1162