Läs mer
4-Methylumbelliferyl 6-Sulfo-2-acetamido-2-deoxy-Alpha-D-glucopyranoside Potassium Salt, TRC
Produktkod. 30474074
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474074 10 mg
30464969 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30474074 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33495010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 210357-37-2
Molekylformel C18 H20 N O11 S . K
Molekylvikt (g/mol) 497.51
Synonym 2H-1-Benzopyran-2-one, 7-[[2-(acetylamino)-2-deoxy-6-O-sulfo-α-D-glucopyranosyl]oxy]-4-methyl-, monopotassium salt (9CI), 4-Methylumbelliferyl 6-sulfo-2-acetamido-2-deoxy-α-D-glucopyranoside potassium salt, 7-[[2-(Acetylamino)-2-deoxy-6-O-sulfo-α-D-glucopyranosyl]oxy]-4-methyl-2H-1-benzopyran-2-one potassium salt, 4-Methylumbelliferyl α-D-N-acetylglucosamine 6-sulfate potassium salt, 4-Methylumbelliferyl 2-acetamido-2-deoxy-α-D-glucopyranoside-6-sulfate potassium salt
IUPAC-namn potassium;[(2R,3S,4R,5R,6R)-5-acetamido-3,4-dihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-2-yl]methyl sulfate
LEDER [K+].CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](COS(=O)(=O)[O-])O[C@@H]1Oc2ccc3C(=CC(=O)Oc3c2)C
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl-6-sulfo-2-acetamido-2-deoxy-beta-D-glucopyranoside potassium salt
Kvantitet 10 mg
InChI formel InChI=1S/C18H21NO11S.K/c1-8-5-14(21)29-12-6-10(3-4-11(8)12)28-18-15(19-9(2)20)17(23)16(22)13(30-18)7-27-31(24,25)26;/h3-6,13,15-18,22-23H,7H2,1-2H3,(H,19,20)(H,24,25,26);/q;+1/p-1/t13-,15-,16-,17-,18+;/m1./s1
Formel vikt 497.0394