Läs mer
4-Methylumbelliferyl 6-Thio-palmitate-Beta-D-glucopyranoside, TRC
Produktkod. 30478954
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478954 50 mg
30475983 5 mg
30466967 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30478954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33667550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 229644-17-1
Molekylformel C32 H48 O8 S
Molekylvikt (g/mol) 592.78
IUPAC-namn S-[[(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-2-yl]methyl] hexadecanethioate
LEDER CCCCCCCCCCCCCCCC(=O)SC[C@H]1O[C@@H](Oc2ccc3C(=CC(=O)Oc3c2)C)[C@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl 6-thio-Palmitate-beta-D-Glucopyranoside
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C32H48O8S/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-28(34)41-21-26-29(35)30(36)31(37)32(40-26)38-23-17-18-24-22(2)19-27(33)39-25(24)20-23/h17-20,26,29-32,35-37H,3-16,21H2,1-2H3/t26-,29-,30+,31-,32-/m1/s1
Formel vikt 592.307
Procent renhet >95%