Läs mer
4-Methylumbelliferyl Alpha-L-Idopyranosiduronic Acid Cyclohexylammonium Salt, TRC
Produktkod. 30135843
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135843 5 mg
30137175 25 mg
17930065 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3347005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66895-33-8
Molekylformel C16 H16 O9 . C6 H13 N
Molekylvikt (g/mol) 451.47
Synonym α-L-Idopyranosiduronic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl, compd. with cyclohexanamine (1:1), 4-Methyl-2-oxo-2H-1-benzopyran-7-yl α-L-idopyranosiduronic acid cyclohexylammonium salt, 4-Methylumbelliferyl α-L-idopyranosiduronic acid cyclohexylammonium salt, 4-Methylumbelliferyl α-L-iduronide cyclohexylammonium salt
IUPAC-namn cyclohexanamine;(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
LEDER CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)ccc12.NC4CCCCC4
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl a-L-Idopyranosiduronic Acid, Cyclohexyl
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C16H16O9.C6H13N/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;7-6-4-2-1-3-5-6/h2-5,11-14,16,18-20H,1H3,(H,21,22);6H,1-5,7H2/t11-,12-,13+,14+,16+;/m0./s1
Formel vikt 451.1842
Procent renhet >95%