Läs mer
4-Methylumbelliferyl Alpha-L-Idopyranosiduronic Acid Methyl Ester, TRC
Produktkod. 30476590
Change view
Kvantitet:
1 mg
2.5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30476590 1 mg -
30476862 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30476590 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3347251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 128095-52-3
Molekylformel C17H18O9
Molekylvikt (g/mol) 366.32
IUPAC-namn methyl (2R,3S,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylate
LEDER COC(=O)[C@@H]1O[C@@H](Oc2ccc3C(=CC(=O)Oc3c2)C)C(O)C(O)[C@@H]1O
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl alpha-L-Idopyranosiduronic Acid Methyl Ester
Kvantitet 1 mg
InChI formel InChI=1S/C17H18O9/c1-7-5-11(18)25-10-6-8(3-4-9(7)10)24-17-14(21)12(19)13(20)15(26-17)16(22)23-2/h3-6,12-15,17,19-21H,1-2H3/t12?,13-,14?,15+,17+/m0/s1
Formel vikt 366.0951