Läs mer
4-Methylumbelliferyl Beta-D-Cellotetroside, TRC
Produktkod. 30481125
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481125 5 mg
30473002 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481125 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3343805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl beta-D-Cellotetroside
CAS 84325-19-9
Molekylformel C34 H48 O22
Kvantitet 5 mg
InChI formel InChI=1S/C34H48O22/c1-11-4-20(40)49-15-5-12(2-3-13(11)15)48-32-27(46)25(44)31(19(10-38)52-32)56-34-28(47)24(43)30(18(9-37)53-34)54-21-6-14(39)29(17(8-36)50-21)55-33-26(45)23(42)22(41)16(7-35)51-33/h2-5,14,16-19,21-39,41-47H,6-10H2,1H3/t14-,16-,17-,18-,19-,21+,22-,23+,24-,25-,26-,27-,28-,29+,30-,31-,32-,33+,34+/m1/s1
LEDER CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O[C@H]5C[C@@H](O)[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@@H](CO)O5)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)ccc12
IUPAC-namn 7-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3,4-dihydroxy-5-[(2S,4R,5S,6R)-4-hydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-methylchromen-2-one
Molekylvikt (g/mol) 808.73
Formel vikt 808.2637