Läs mer
4-Methylumbelliferyl Beta-D-N,N’,N' '-Triacetylchitotrioside, TRC
Produktkod. 30138650
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138650 10 mg
30138243 1 mg
30012428 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138650 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33750010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53643-13-3
Molekylformel C34 H47 N3 O18
Molekylvikt (g/mol) 785.75
IUPAC-namn N-[(2S,3R,4R,5S,6R)-2-[(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,3S,4R,5R,6S)-5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
LEDER CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1O[C@H]2[C@H](O)[C@@H](NC(=O)C)[C@H](O[C@H]3[C@H](O)[C@@H](NC(=O)C)[C@H](Oc4ccc5C(=CC(=O)Oc5c4)C)O[C@@H]3CO)O[C@@H]2CO
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl beta-D-N,N',N' '-Triacetylchitotrioside
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C34H47N3O18/c1-12-7-22(44)50-18-8-16(5-6-17(12)18)49-32-24(36-14(3)42)28(47)30(20(10-39)52-32)55-34-25(37-15(4)43)29(48)31(21(11-40)53-34)54-33-23(35-13(2)41)27(46)26(45)19(9-38)51-33/h5-8,19-21,23-34,38-40,45-48H,9-11H2,1-4H3,(H,35,41)(H,36,42)(H,37,43)/t19-,20-,21-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33+,34+/m1/s1
Formel vikt 785.2855
Procent renhet >95%