Läs mer
Melford 4-Methylumbellifery-α -D-mannopyranosid
Produktkod. 30018529
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30018529 100 mg
30018528 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30018529 Leverantör Melford Leverantörsnummer M666000.1
 Begränsad artikel
Beräknad leverans: 28-10-2026
Fluorescerande substrat förα -D-Mannosidas.
Fluorescerande substrat förα -D-Mannosidas. 4-metylumbelliferon:< 50 ppm.