Läs mer
4-Methylumbelliferyl Oleate, TRC
Produktkod. 30474232
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474232 100 mg
30476642 50 mg
30465161 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474232 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM334775100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18323-58-5
Molekylformel C28 H40 O4
Molekylvikt (g/mol) 440.61
Synonym 9-Octadecenoic acid (9Z)-, 4-methyl-2-oxo-2H-1-benzopyran-7-yl ester, 9-Octadecenoic acid (Z)-, 4-methyl-2-oxo-2H-1-benzopyran-7-yl ester, Oleic acid, ester with 7-hydroxy-4-methylcoumarin (8CI), Coumarin, 7-hydroxy-4-methyl-, oleate (8CI), 4-Methylumbelliferone oleate, 4-Methylumbelliferyl oleate, Oleoyl 4-methylumbelliferone
IUPAC-namn (4-methyl-2-oxochromen-7-yl) (Z)-octadec-9-enoate
LEDER CCCCCCCC\C=C/CCCCCCCC(=O)Oc1ccc2C(=CC(=O)Oc2c1)C
Specifikationer
Kemiskt namn eller material 4-Methylumbelliferyl Oleate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C28H40O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-27(29)31-24-19-20-25-23(2)21-28(30)32-26(25)22-24/h10-11,19-22H,3-9,12-18H2,1-2H3/b11-10-
Formel vikt 440.2927
Procent renhet >95%