missing translation for 'onlineSavingsMsg'
Läs mer

4-Methylumbelliferyl Oleate, TRC

Produktkod. 30476642
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476642 50 mg
30474232 100 mg
30465161 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30476642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33477550MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 18323-58-5
Molekylformel C28 H40 O4
Molekylvikt (g/mol) 440.61
Synonym 9-Octadecenoic acid (9Z)-, 4-methyl-2-oxo-2H-1-benzopyran-7-yl ester, 9-Octadecenoic acid (Z)-, 4-methyl-2-oxo-2H-1-benzopyran-7-yl ester, Oleic acid, ester with 7-hydroxy-4-methylcoumarin (8CI), Coumarin, 7-hydroxy-4-methyl-, oleate (8CI), 4-Methylumbelliferone oleate, 4-Methylumbelliferyl oleate, Oleoyl 4-methylumbelliferone
IUPAC-namn (4-methyl-2-oxochromen-7-yl) (Z)-octadec-9-enoate
LEDER CCCCCCCC\C=C/CCCCCCCC(=O)Oc1ccc2C(=CC(=O)Oc2c1)C

Specifikationer

Kemiskt namn eller material 4-Methylumbelliferyl Oleate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C28H40O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-27(29)31-24-19-20-25-23(2)21-28(30)32-26(25)22-24/h10-11,19-22H,3-9,12-18H2,1-2H3/b11-10-
Formel vikt 440.2927
Procent renhet >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.