Läs mer
4-Morpholineethanesulfonic Acid, TRC
Produktkod. 30479928
Change view
Kvantitet:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479928 100 g
30481763 50 g
30466732 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479928 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM723775100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4432-31-9
Molekylformel C6 H13 N O4 S
Molekylvikt (g/mol) 195.24
Synonym 3-(N-Morpholino)ethane Sulfonic Acid, 4-Morpholineethanesulfonic acid, 2-(4-Morpholino)ethanesulfonic acid, 2-(4-Morpholinyl)ethanesulfonic acid, 2-(N-Morpholino)ethanesulfonic acid, 2-Morpholinoethanesulfonic acid, MES, MES (buffering agent), Morpholinoethanesulfonic acid, NSC 157116
IUPAC-namn 2-morpholin-4-ylethanesulfonic acid
LEDER OS(=O)(=O)CCN1CCOCC1
Specifikationer
Kemiskt namn eller material 3-(N-Morpholino)ethane Sulfonic Acid (MES)
Kvantitet 100 g
InChI formel InChI=1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)
Formel vikt 195.0565