missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)benzoic acid, Maybridge™

360.41 SEK - 5569.14 SEK

Chemical Identifiers

CAS 62642-62-0
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
InChI Key QYBXZYYECZFQRX-UHFFFAOYSA-N
Synonym 4-morpholinomethyl benzoic acid, 4-morpholin-4-ylmethyl benzoic acid, 4-morpholin-4-ylmethyl-benzoic acid, 4-4-morpholinemethyl benzoic acid, 4-4-morpholinylmethyl benzoic acid, benzoic acid, 4-4-morpholinylmethyl, timtec-bb sbb011465, cbmicro_000050, acmc-1b2v0, 4-morpholinomethylbenzoic acid
PubChem CID 703507
IUPAC Name 4-(morpholin-4-ylmethyl)benzoic acid
SMILES C1COCCN1CC2=CC=C(C=C2)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10127892
SDS View Documents
Maybridge
CC29301CB
250mg Amber Glass Bottle
360.41 SEK
250mg
View alternative products
Save Up to 
»
Estimated Shipment
17-11-2020
Add to basket
 
10012172
SDS View Documents
Maybridge
CC29301DA
1g Amber Glass Bottle
810.92 SEK
1g
View alternative products
Save Up to 
»
Estimated Shipment
17-11-2020
Add to basket
 
10245474
SDS View Documents
Maybridge
CC29301EA
10g Amber Glass Bottle
5569.14 SEK
10g
View alternative products
Save Up to 
»
Estimated Shipment
17-11-2020
Add to basket
 
Specifications

Chemical Identifiers

62642-62-0
221.256
4-morpholinomethyl benzoic acid, 4-morpholin-4-ylmethyl benzoic acid, 4-morpholin-4-ylmethyl-benzoic acid, 4-4-morpholinemethyl benzoic acid, 4-4-morpholinylmethyl benzoic acid, benzoic acid, 4-4-morpholinylmethyl, timtec-bb sbb011465, cbmicro_000050, acmc-1b2v0, 4-morpholinomethylbenzoic acid
4-(morpholin-4-ylmethyl)benzoic acid
C12H15NO3
QYBXZYYECZFQRX-UHFFFAOYSA-N
703507
C1COCCN1CC2=CC=C(C=C2)C(=O)O

Specifications

4-(Morpholinomethyl)benzoic acid
250mg
97%
4-morpholinomethyl benzoic acid, 4-morpholin-4-ylmethyl benzoic acid, 4-morpholin-4-ylmethyl-benzoic acid, 4-4-morpholinemethyl benzoic acid, 4-4-morpholinylmethyl benzoic acid, benzoic acid, 4-4-morpholinylmethyl, timtec-bb sbb011465, cbmicro_000050, acmc-1b2v0, 4-morpholinomethylbenzoic acid
C1COCCN1CC2=CC=C(C=C2)C(=O)O
221.256
221.25
Amber Glass Bottle
62642-62-0
C12H15NO3
QYBXZYYECZFQRX-UHFFFAOYSA-N
4-(morpholin-4-ylmethyl)benzoic acid
703507
97%
SDS