missing translation for 'onlineSavingsMsg'
Learn More

4-(N-[2-(Dimethylamino)ethyl]carbamoyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7034583
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
832114-11-1
C17H27BN2O3
318.224
MFCD05864307
KTDBEJMIGCLCSU-UHFFFAOYSA-N
n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-n',n'-dimethylamino ethyl benzamide-4-boronic acid, pinacol ester, n-2-dimethylamino ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylamino ethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-dimethylamino ethylcarbamoyl phenylboronic acid, pinacol ester, amtb351, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,, 4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester, 4-n-2-dimethylamino ethyl carbamoyl benzeneboronic acid pinacol ester
2758503
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C
This item is not returnable. View return policy
832114-11-1
C17H27BN2O3
318.224
MFCD05864307
KTDBEJMIGCLCSU-UHFFFAOYSA-N
n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-n',n'-dimethylamino ethyl benzamide-4-boronic acid, pinacol ester, n-2-dimethylamino ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylamino ethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-dimethylamino ethylcarbamoyl phenylboronic acid, pinacol ester, amtb351, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,, 4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester, 4-n-2-dimethylamino ethyl carbamoyl benzeneboronic acid pinacol ester
2758503
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 318.22
Percent Purity 95%
Chemical Name or Material 4-(N-[2-(Dimethylamino)ethyl]carbamoyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'