missing translation for 'onlineSavingsMsg'
Learn More

4-N-BOC-4-N-methylaminopiperidine, 98%, ACROS Organics™

Brand:  Acros Organics 455890050

Additional Details : CAS Number : 108612-54-0 Weight : 0.00500kg


Quantity 5g

 View more versions of this product

Product Code. 15565906

  • 5087.00 SEK / 5g


Estimated Shipment
17-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

108612-54-0
214.31
tert-butyl methyl piperidin-4-yl carbamate, 4-n-boc-4-n-methyl-aminopiperidine, 4-n-boc-4-n-methylaminopiperidine, tert-butyl n-methyl-n-piperidin-4-yl carbamate, 4-n-boc-n-methylamino piperidine, 4-boc-4-methylaminopiperidine, methyl-piperidin-4-yl-carbamic acid tert-butyl ester, tert-butyl n-methyl-n-4-piperidyl carbamate, tert-butyl methylpiperidin-4-ylcarbamate, methyl-piperidine-4-yl-carbamic acid tert-butyl ester
tert-butyl N-methyl-N-piperidin-4-ylcarbamate
C11H22N2O2
DJJOYDXRUBOZON-UHFFFAOYSA-N
2756806
CC(C)(C)OC(=O)N(C)C1CCNCC1

Specifications

4-N-BOC-4-N-methylaminopiperidine
39.0°C to 41.0°C
108612-54-0
C11H22N2O2
DJJOYDXRUBOZON-UHFFFAOYSA-N
tert-butyl N-methyl-N-piperidin-4-ylcarbamate
2756806
98%
Authentic
5g
97.5% min. (GC)
tert-butyl methyl piperidin-4-yl carbamate, 4-n-boc-4-n-methyl-aminopiperidine, 4-n-boc-4-n-methylaminopiperidine, tert-butyl n-methyl-n-piperidin-4-yl carbamate, 4-n-boc-n-methylamino piperidine, 4-boc-4-methylaminopiperidine, methyl-piperidin-4-yl-carbamic acid tert-butyl ester, tert-butyl n-methyl-n-4-piperidyl carbamate, tert-butyl methylpiperidin-4-ylcarbamate, methyl-piperidine-4-yl-carbamic acid tert-butyl ester
CC(C)(C)OC(=O)N(C)C1CCNCC1
214.31
214.31
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-N-Boc-4-N-Methyl-aminopiperidine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents