Learn More
4-n-Butoxy-2,6-difluoroaniline, 97%, Thermo Scientific™
Catalog Number 15495927
Click to view available options
Quantity:
250 mg
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
CAS 1373920-83-2
Molecular Formula C10H13F2NO
MDL Number MFCD22201028
Quantity 250 mg
UN Number UN2810
Synonym 4-n-butoxy-2,6-difluoroaniline
InChI Key QJBXYVXNGAAECC-UHFFFAOYSA-N
SMILES CCCCOC1=CC(F)=C(N)C(F)=C1
IUPAC Name 4-butoxy-2,6-difluoroaniline
Molecular Weight (g/mol) 201.22
PubChem CID 86277631
Formula Weight 201.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-n-Butoxy-2,6-difluoroaniline