missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxy-2,6-difluorobenzonitrile, 97%, Thermo Scientific™

Catalog Number p-7052414
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1373920-94-5
C11H11F2NO
211.212
MFCD22201026
DRMAQHPUVUFLAE-UHFFFAOYSA-N
4-n-butoxy-2,6-difluorobenzonitrile
86277629
4-butoxy-2,6-difluorobenzonitrile
CCCCOC1=CC(=C(C(=C1)F)C#N)F
This item is not returnable. View return policy
1373920-94-5
C11H11F2NO
211.212
MFCD22201026
DRMAQHPUVUFLAE-UHFFFAOYSA-N
4-n-butoxy-2,6-difluorobenzonitrile
86277629
4-butoxy-2,6-difluorobenzonitrile
CCCCOC1=CC(=C(C(=C1)F)C#N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 211.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-n-Butoxy-2,6-difluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'