missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Decyloxybenzoic acid, 96%, Thermo Scientific Chemicals
323.46 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,
Solubility
Soluble in water.
Notes
m.p. 97°(S), 122°(N), 142°(I)
4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,
Solubility
Soluble in water.
Notes
m.p. 97°(S), 122°(N), 142°(I)
Specifications
Melting Point | 97°C (S) |
Quantity | 5 g |
Beilstein | 2113730 |
Synonym | 4-decyloxy benzoic acid, p-decyloxybenzoic acid, 4-n-decyloxybenzoic acid, 4-decyloxybenzoic acid, benzoic acid,4-decyloxy, benzoic acid, 4-decyloxy, pubchem2671, p-n-decyloxybenzoic acid, 4-decyloxy benzoicacid, para-decyloxy benzoic acid |
Solubility Information | Soluble in water. |
Formula Weight | 278.39 |
Percent Purity | 96% |
Chemical Name or Material | 4-n-Decyloxybenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'