missing translation for 'onlineSavingsMsg'
Learn More

4-n-Dodecylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7031049
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
21021-55-6
C19H30O2
290.447
MFCD04972235
JCHCIOJFCCQEKZ-UHFFFAOYSA-N
p-dodecylbenzoic acid, benzoic acid, 4-dodecyl, p-n-dodecyl benzoic acid, benzoicacid,4-dodecyl, 4-n-dodecylbenzoic acid
413311
4-dodecylbenzoic acid
CCCCCCCCCCCCC1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
21021-55-6
C19H30O2
290.447
MFCD04972235
JCHCIOJFCCQEKZ-UHFFFAOYSA-N
p-dodecylbenzoic acid, benzoic acid, 4-dodecyl, p-n-dodecyl benzoic acid, benzoicacid,4-dodecyl, 4-n-dodecylbenzoic acid
413311
4-dodecylbenzoic acid
CCCCCCCCCCCCC1=CC=C(C=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 290.45
Percent Purity 98%
Chemical Name or Material 4-n-Dodecylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'