Läs mer
4-(N,N-Dimethylamino)butanal Dimethyl Acetal, TRC
Produktkod. 30369198
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30369198 25 g
1 options
Produktkod. 30369198 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD46140525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(N,N-Dimethylamino)butanal Dimethyl Acetal
CAS 19718-92-4
Molekylformel C8H19NO2
Kvantitet 25 g
Synonym 1-Butanamine, 4,4-dimethoxy-N,N-dimethyl-, Butyraldehyde, 4-(dimethylamino)-, dimethyl acetal (8CI), 4,4-Dimethoxy-N,N-dimethyl-1-butanamine, Butylamine, 4,4-dimethoxy-N,N-dimethyl- (8CI), 4,4-Dimethoxy-N,N-dimethylbutanamine, 4-(Dimethylamino)butanal dimethyl acetal, 4-(Dimethylamino)butyraldehyde dimethyl acetal, 4-(Dimethylamino)butyraldehyde Dimethyl Acetal, 4,4-Dimethoxy-N,N-dimethylbutanamine, 4,4-Dimethoxy-N,N-dimethylbutan-1-amine
InChI formel InChI=1S/C8H19NO2/c1-9(2)7-5-6-8(10-3)11-4/h8H,5-7H2,1-4H3
LEDER COC(CCCN(C)C)OC
IUPAC-namn 4,4-dimethoxy-N,N-dimethylbutan-1-amine
Molekylvikt (g/mol) 161.24
Formel vikt 161.1416