Läs mer
4-N-Propylbenzaldehyde, TRC
Produktkod. 30428029
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428029 1 g
30424496 500 mg
30411260 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428029 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8353181G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28785-06-0
Molekylformel C10 H12 O
Molekylvikt (g/mol) 148.2017
Synonym 4-n-Propylbenzaldehyde, Benzaldehyde, p-propyl- (8CI), 4-Propylbenzaldehyde, 4-(n-Propyl)benzaldehyde, [4-Propylphenyl]methanone, p-Propylbenzaldehyde, Benzaldehyde, 4-propyl-
IUPAC-namn 4-propylbenzaldehyde
LEDER CCCc1ccc(C=O)cc1
Specifikationer
Kemiskt namn eller material 4-n-Propylbenzaldehyde
Kvantitet 1 g
InChI formel InChI=1S/C10H12O/c1-2-3-9-4-6-10(8-11)7-5-9/h4-8H,2-3H2,1H3
Formel vikt 148.0888