missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2-(trifluoromethyl)benzoxazole, 97%, Thermo Scientific™

Catalog Number p-7055106
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
573759-00-9
C8H3F3N2O3
232.118
MVYGNYLHXPFQJV-UHFFFAOYSA-N
4-nitro-2-trifluoromethyl benzo d oxazole, 4-nitro-2-trifluoromethylbenzoxazole, 4-nitro-2-trifluoromethyl-1,3-benzoxazole, 4-nitro-2-trifluoromethyl benzoxazole, 4-nitro-2-trifluoromethyl-benzooxazole
21875019
4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
C1=CC(=C2C(=C1)OC(=N2)C(F)(F)F)[N+](=O)[O-]
This item is not returnable. View return policy
573759-00-9
C8H3F3N2O3
232.118
MVYGNYLHXPFQJV-UHFFFAOYSA-N
4-nitro-2-trifluoromethyl benzo d oxazole, 4-nitro-2-trifluoromethylbenzoxazole, 4-nitro-2-trifluoromethyl-1,3-benzoxazole, 4-nitro-2-trifluoromethyl benzoxazole, 4-nitro-2-trifluoromethyl-benzooxazole
21875019
4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
C1=CC(=C2C(=C1)OC(=N2)C(F)(F)F)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 232.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)benzoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'