missing translation for 'onlineSavingsMsg'
Läs mer

4-Nitroaniline, Sigma-Aldrich™

Produktkod. 15871761
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 g
1 kg
Förpackningsstorlek:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15871761 1 kg 1kg
15861761 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15871761 Leverantör Sigma Aldrich Leverantörsnummer 8222921000
 Begränsad artikel
Beräknad leverans: 28-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Kvalitet Research
Molekylvikt (g/mol) 138.12

Specifications

Smältpunkt 148°C
Densitet 1.44 g/cm3 at 20°C
Kokpunkt >331°C/1013 hPa (Decomposition)
MDL-nummer MFCD00007858
Kvantitet 1 kg
Synonym 4-Nitroaniline, p-Nitraniline, 4-Nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.