missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number 11428427
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
98-74-8
C6H4ClNO4S
221.61
MFCD00007445
JXRGUPLJCCDGKG-UHFFFAOYSA-N
4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
7404
[O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
98-74-8
C6H4ClNO4S
221.61
MFCD00007445
JXRGUPLJCCDGKG-UHFFFAOYSA-N
4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
7404
[O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O

4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2'-bipyridine, nbs = 4-nitrobenzenesulfonate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2′-bipyridine, nbs = 4-nitrobenzenesulfonate).

Solubility
Soluble in toluene, tetrahydrofuran, methylene chloride and ethyl acetate. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 76°C to 80°C
Boiling Point 143°C to 144°C (1.5 mmHg)
Quantity 100 g
UN Number UN3261
Beilstein 746543
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene,tetrahydrofuran,methylene chloride and ethyl acetate. Insoluble in water.
IUPAC Name 4-nitrobenzene-1-sulfonyl chloride
Formula Weight 221.62
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'