missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzo-15-crown-5, 99%, Thermo Scientific Chemicals

Catalog Number p-7023138
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
60835-69-0
C14H19NO7
313.306
MFCD00060713
XIWRBQVYCZCEPG-UHFFFAOYSA-N
4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether
143747
17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene
C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1
This item is not returnable. View return policy
60835-69-0
C14H19NO7
313.306
MFCD00060713
XIWRBQVYCZCEPG-UHFFFAOYSA-N
4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether
143747
17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene
C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1

4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.

Solubility
It is soluble in non-polar solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases
TRUSTED_SUSTAINABILITY
Melting Point 93°C to 95°C
Quantity 1 g
Solubility Information It is soluble in non-polar solvents.
Formula Weight 313.3
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzo-15-crown-5

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'