Läs mer
4-Nitrobenzyl Acetoacetate, TRC
Produktkod. 30482597
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30482597 10 g
1 options
Produktkod. 30482597 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49543010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Nitrobenzyl Acetoacetate
CAS 61312-84-3
Molekylformel C11H11NO5
Kvantitet 10 g
Synonym 4-Nitrobenzyl Acetylacetate, p-Nitrobenzyl 3-Oxobutanoate, p-Nitrobenzyl Acetoacetate
InChI formel InChI=1S/C11H11NO5/c1-8(13)6-11(14)17-7-9-2-4-10(5-3-9)12(15)16/h2-5H,6-7H2,1H3
LEDER CC(=O)CC(=O)OCc1ccc(cc1)[N+](=O)[O-]
IUPAC-namn (4-nitrophenyl)methyl 3-oxobutanoate
Molekylvikt (g/mol) 237.21
Formel vikt 237.0637