missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzyl alcohol, 99%, Thermo Scientific Chemicals
302.96 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.
Solubility
Soluble in water (2 mg/ml at 20°C).
Notes
Store at room temperature. Keep away from oxidizing agents.
4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.
Solubility
Soluble in water (2 mg/ml at 20°C).
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Melting Point | 92°C to 95°C |
Boiling Point | 185°C (12 mmHg) |
Flash Point | 180°C (356°F) |
Quantity | 25 g |
Beilstein | 1424026 |
Solubility Information | Soluble in water (2mg/ml at 20°C). |
Formula Weight | 153.14 |
Percent Purity | 99% |
Chemical Name or Material | 4-Nitrobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'