missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzyl bromide, 97+%, Thermo Scientific Chemicals
946.74 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.
Solubility
It hydrolyzes in water. Soluble in alcohol and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.
Solubility
It hydrolyzes in water. Soluble in alcohol and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
Specifications
Melting Point | 97°C to 101°C |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 742796 |
Solubility Information | It hydrolyzes in water. Soluble in alcohol and ether. |
Formula Weight | 216.04 |
Percent Purity | ≥97% |
Chemical Name or Material | 4-Nitrobenzyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'