missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl bromide, 97+%, Thermo Scientific Chemicals

Catalog Number p-7022448
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
100-11-8
C7H6BrNO2
216.03
MFCD00007373
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
66011
CHEBI:62822
1-(bromomethyl)-4-nitrobenzene
[O-][N+](=O)C1=CC=C(CBr)C=C1
This item is not returnable. View return policy
100-11-8
C7H6BrNO2
216.03
MFCD00007373
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
66011
CHEBI:62822
1-(bromomethyl)-4-nitrobenzene
[O-][N+](=O)C1=CC=C(CBr)C=C1

4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

Solubility
It hydrolyzes in water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
TRUSTED_SUSTAINABILITY
Melting Point 97°C to 101°C
Quantity 25 g
UN Number UN3261
Beilstein 742796
Solubility Information It hydrolyzes in water. Soluble in alcohol and ether.
Formula Weight 216.04
Percent Purity ≥97%
Chemical Name or Material 4-Nitrobenzyl bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'