Läs mer
4-Nitrophenyl 2,3,4-tri-O-acetyl-Beta-D-glucuronic Acid, Methyl Ester, TRC
Produktkod. 30477366
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30477366 100 mg
1 options
Produktkod. 30477366 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN506750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Nitrophenyl 2,3,4-tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester
CAS 18472-49-6
Molekylformel C19 H21 N O12
Kvantitet 100 mg
Synonym 4-Nitrophenyl 2,3,4-Tri-O-Acetyl-B-D-Glucuronic Acid, Methyl Ester
InChI formel InChI=1S/C19H21NO12/c1-9(21)28-14-15(29-10(2)22)17(30-11(3)23)19(32-16(14)18(24)27-4)31-13-7-5-12(6-8-13)20(25)26/h5-8,14-17,19H,1-4H3/t14-,15-,16-,17+,19+/m1/s1
LEDER COC(=O)[C@@H]1O[C@H](Oc2ccc(cc2)[N+](=O)[O-])[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC-namn methyl (2R,3R,4R,5S,6R)-3,4,5-triacetyloxy-6-(4-nitrophenoxy)oxane-2-carboxylate
Molekylvikt (g/mol) 455.37
Formel vikt 455.1064