Läs mer
4-Nitrophenyl 2,4,6-Tri-O-acetyl-3-O-(2,3,4,6-tetra-O-acetyl -Beta-D-glucopyranosyl)-b-D-glucopyranoside, TRC
Produktkod. 30464441
Change view
Kvantitet:
2 mg
Produktkod. Kvantitet
30464441 2 mg
1 options
Produktkod. 30464441 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5069252MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Nitrophenyl 2,4,6-Tri-O-acetyl-3-O-(2,3,4,6-tetra-O-acetyl -beta-D-glucopyranosyl)-b-D-glucopyranoside
CAS 195715-98-1
Molekylformel C32H39NO20
Kvantitet 2 mg
Synonym 4-Nitrophenyl 3-O-(2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl)-beta-D-glucopyranoside 2,4,6-Triacetate
InChI formel InChI=1S/C32H39NO20/c1-14(34)43-12-23-25(45-16(3)36)27(47-18(5)38)29(48-19(6)39)32(52-23)53-28-26(46-17(4)37)24(13-44-15(2)35)51-31(30(28)49-20(7)40)50-22-10-8-21(9-11-22)33(41)42/h8-11,23-32H,12-13H2,1-7H3/t23-,24-,25-,26-,27+,28+,29-,30-,31-,32+/m1/s1
LEDER CC(=O)OC[C@H]1O[C@@H](O[C@H]2[C@H](OC(=O)C)[C@@H](COC(=O)C)O[C@@H](Oc3ccc(cc3)[N+](=O)[O-])[C@@H]2OC(=O)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn [(2R,3R,4S,5R,6S)-3,5-diacetyloxy-6-(4-nitrophenoxy)-4-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
Molekylvikt (g/mol) 757.65
Formel vikt 757.2065