Läs mer
4-Nitrophenyl-Alpha-L-arabinofuranoside, TRC
Produktkod. 30481115
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481115 25 mg
30480359 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481115 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50295025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Nitrophenyl-alpha-L-arabinofuranoside
CAS 6892-58-6
Molekylformel C11 H13 N O7
Kvantitet 25 mg
Synonym p-Nitrophenylarabinofuranoside, p-Nitrophenyl-α-L-arabinofuranoside
InChI formel InChI=1S/C11H13NO7/c13-5-8-9(14)10(15)11(19-8)18-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2/t8-,9-,10+,11+/m0/s1
LEDER OC[C@@H]1O[C@@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](O)[C@H]1O
IUPAC-namn (2S,3R,4R,5S)-2-(hydroxymethyl)-5-(4-nitrophenoxy)oxolane-3,4-diol
Molekylvikt (g/mol) 271.22
Formel vikt 271.0692