missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl formate, 98%, Thermo Scientific Chemicals

Catalog Number p-7043341
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1865-01-6
C7H5NO4
167.12
MFCD00014713
IEXRKQFZXJSHOB-UHFFFAOYSA-N
74628
(4-nitrophenyl) formate
C1=CC(=CC=C1[N+](=O)[O-])OC=O
This item is not returnable. View return policy
1865-01-6
C7H5NO4
167.12
MFCD00014713
IEXRKQFZXJSHOB-UHFFFAOYSA-N
74628
(4-nitrophenyl) formate
C1=CC(=CC=C1[N+](=O)[O-])OC=O

Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

Solubility
Hydrolyzes in water. Soluble in dichloromethane.

Notes
Moisture and light sensitive. Store away from water/moisture, oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark. Store under inert gas. Protect from exposure to light.
TRUSTED_SUSTAINABILITY
Melting Point 70°C to 74°C
Quantity 5 g
Beilstein 1950897
Sensitivity Light and moisture sensitive
Synonym p-nitrophenyl formate, formic acid, 4-nitrophenyl ester, 4-nitrophenyl formate, formic acid, p-nitrophenyl ester, formic acid 4-nitrophenyl ester, acmc-1bty1, formic acid,4-nitrophenyl ester, formic acid 4-nitro-phenyl ester, 4-nitrophenyl formate hplc
Solubility Information Hydrolyzes in water. Soluble in dichloromethane.
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 4-Nitrophenyl formate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'