Läs mer
4-Nitrophenyl Isocyanide, TRC
Produktkod. 30464668
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30464668 50 mg
1 options
Produktkod. 30464668 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN54528850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Nitrophenyl Isocyanide
CAS 1984-23-2
Molekylformel C7 H4 N2 O2
Kvantitet 50 mg
Synonym p-Nitrophenyl Isocyanide, p-Nitrophenyl Isonitrile, 1-Isocyano-4-nitrobenzene
InChI formel InChI=1S/C7H4N2O2/c1-8-6-2-4-7(5-3-6)9(10)11/h2-5H
LEDER [O-][N+](=O)c1ccc(cc1)[N+]#[C-]
IUPAC-namn 1-isocyano-4-nitrobenzene
Molekylvikt (g/mol) 148.119
Formel vikt 148.027