missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
1885.60 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 Pc.
1000 Pc.
Description
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.
Solubility
It is soluble in water.
Notes
The tablets should be stored -20°C
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.
Solubility
It is soluble in water.
Notes
The tablets should be stored -20°C
Specifications
Color | Yellow |
Quantity | 100 Pc. |
Beilstein | 5324661 |
Solubility Information | It is soluble in water. |
Formula Weight | 371.15 (263.05 Anhydrous) |
Physical Form | Tablets |
Chemical Name or Material | 4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'