Läs mer
4-Nitrophenyl Propionate, TRC
Produktkod. 30418380
Change view
Kvantitet:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418380 25 mg
30424595 50 mg
30423699 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30418380 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN92617825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19560-60-5
Molekylformel C9 H9 N O4
Molekylvikt (g/mol) 195.172
IUPAC-namn (4-nitrophenyl) propanoate
LEDER CCC(=O)Oc1ccc(cc1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 4-Nitrophenyl Propionate
Kvantitet 25 mg
InChI formel InChI=1S/C9H9NO4/c1-2-9(11)14-8-5-3-7(4-6-8)10(12)13/h3-6H,2H2,1H3
Formel vikt 195.053