Läs mer
4-Nitrophenyl Thiazol-5-ylmethyl Carbonate, Mikromol™
Produktkod. 30459305
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30459305 4 x 25 mg
30049105 25 mg
17907134 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30459305 Leverantör Mikromol Leverantörsnummer MM1034.2300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Nitrophenyl Thiazol-5-ylmethyl Carbonate, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Thiazolylmethyl-4-nitrophenylcarbonate
CAS 144163-97-3
Analyt- eller komponentnamn Thiazolylmethyl-4-nitrophenylcarbonate
Frakt skick Room Temperature
Molekylformel C11 H8 N2 O5 S
Certifikat och efterlevnad ISO 9001
Synonym Carbonic acid, 4-nitrophenyl 5-thiazolylmethyl ester, (4-Nitrophenyl) thiazol-5-ylmethyl carbonate, Thiazolylmethyl-4-nitrophenylcarbonate
InChI formel InChI=1S/C11H8N2O5S/c14-11(17-6-10-5-12-7-19-10)18-9-3-1-8(2-4-9)13(15)16/h1-5,7H,6H2
Lösningstyp Neat
LEDER [O-][N+](=O)c1ccc(OC(=O)OCc2cncs2)cc1
IUPAC-namn (4-nitrophenyl) 1,3-thiazol-5-ylmethyl carbonate
Molekylvikt (g/mol) 280.26
Kvantitet 4 x 25 mg
Formel vikt 280.0154 g/mol