Läs mer
4-Nitrophenyl trifluoroacetate, TRC
Produktkod. 30477181
Change view
Kvantitet:
1 g
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30477181 1 g -
30468636 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30477181 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN2326051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Nitrophenyl trifluoroacetate
CAS 658-78-6
Molekylformel CF3CO2C6H4NO2
Kvantitet 1 g
InChI formel InChI=1S/C8H4F3NO4/c9-8(10,11)7(13)16-6-3-1-5(2-4-6)12(14)15/h1-4H
LEDER [O-][N+](=O)c1ccc(OC(=O)C(F)(F)F)cc1
IUPAC-namn (4-nitrophenyl) 2,2,2-trifluoroacetate
Molekylvikt (g/mol) 235.12
Formel vikt 235.0092