missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophthalic anhydride, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
4-Nitrophthalic anhydride is used an an intermediate for the synthesis of a benzimidazole PARP inhibitor I (succinate salt) (ABT-472).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophthalic anhydride is used an an intermediate for the synthesis of a benzimidazole PARP inhibitor I (succinate salt) (ABT-472).
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases, strong acids and oxidizing agents.
4-Nitrophthalic anhydride is used an an intermediate for the synthesis of a benzimidazole PARP inhibitor I (succinate salt) (ABT-472).
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases, strong acids and oxidizing agents.
Specifications
Melting Point | 116°C to 121°C |
Quantity | 5 g |
Beilstein | 179682 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 193.11 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | 4-Nitrophthalic anhydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'