Läs mer
4-Nitroso Sulfamethoxazole, >90%, TRC
Produktkod. 30483166
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30483166 25 mg -
30468467 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483166 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN54670025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 131549-85-4
Molekylformel C10 H9 N3 O4 S
Molekylvikt (g/mol) 267.26
Synonym N-(5-Methyl-3-isoxazolyl)-4-nitrosobenzenesulfonamide, 4-Nitrososulfamethoxazole
IUPAC-namn N-(5-methyl-1,2-oxazol-3-yl)-4-nitrosobenzenesulfonamide
LEDER Cc1onc(NS(=O)(=O)c2ccc(cc2)N=O)c1
Specifikationer
Kemiskt namn eller material 4-Nitroso Sulfamethoxazole
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H9N3O4S/c1-7-6-10(12-17-7)13-18(15,16)9-4-2-8(11-14)3-5-9/h2-6H,1H3,(H,12,13)
Formel vikt 267.0314
Procent renhet >95%