Läs mer
4-Nonyl Phenol-13C6 Diethoxylate, TRC
Produktkod. 30472296
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30472296 5 mg
1 options
Produktkod. 30472296 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN6504425MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-n-Nonylphenol diethoxylate ring-13C6
CAS 1346602-11-6
Molekylformel 13C6 C13 H32 O3
Kvantitet 5 mg
Synonym 2-[2-(4-nonylphenoxy)ethoxy]ethanol-13C6, 2-[2-(p-Nonylphenoxy)ethoxy]ethanol-13C6, 4-n-Nonylphenol Diethoxylate-13C6, Diethylene Glycol Mono-p-nonylphenyl Ether-13C6, Diethylene Glycol p-Nonylphenyl Ether-13C6, EON 2-13C6
InChI formel InChI=1S/C19H32O3/c1-2-3-4-5-6-7-8-9-18-10-12-19(13-11-18)22-17-16-21-15-14-20/h10-13,20H,2-9,14-17H2,1H3/i10+1,11+1,12+1,13+1,18+1,19+1
LEDER CCCCCCCCC[13c]1[13cH][13cH][13c](OCCOCCO)[13cH][13cH]1
IUPAC-namn 2-[2-(4-nonyl(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-yl)oxyethoxy]ethanol
Molekylvikt (g/mol) 314.41
Formel vikt 314.255