Läs mer
4-Nonyl Phenol-13C6, TRC
Produktkod. 30010189
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010189 1 mg
30003998 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010189 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN6504321MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 211947-56-7
Molekylformel 13C6 C9 H24 O
Molekylvikt (g/mol) 226.31
Synonym p-Nonylhenol-13C6, 4-Nonylphenol-13C6, 4-n-Nonyl Phenol-13C6, p-NP-13C6, p-Nonylphenol-13C6, p-n-Nonylphenol-13C6
IUPAC-namn 4-nonyl(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-ol
LEDER CCCCCCCCC[13c]1[13cH][13cH][13c](O)[13cH][13cH]1
Specifikationer
Kemiskt namn eller material p-n-Nonylphenol ring-13C6
Kvantitet 1 mg
InChI formel InChI=1S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16H,2-9H2,1H3/i10+1,11+1,12+1,13+1,14+1,15+1
Formel vikt 226.203