Läs mer
4'-O-Demethyl-pantoprazole sulfide, TRC
Produktkod. 30011585
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011585 25 mg -
30008929 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011585 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23240025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 141854-21-9
Molekylformel C15H13F2N3O3S
Molekylvikt (g/mol) 353.34
Synonym 2-[[[6-(Difluoromethoxy)-1H-benzimidazol-2-yl]thio]methyl]-3-methoxy-4-pyridinol4'-O-Demethyl-pantopraz, 4'-O-Demethyl-pantoprazole thioether
LEDER COc1c(O)ccnc1CSc2nc3ccc(OC(F)F)cc3[nH]2
Specifikationer
Kemiskt namn eller material 4'-O-Demethyl-pantoprazole sulfide
Kvantitet 25 mg
InChI formel InChI=1S/C15H13F2N3O3S/c1-22-13-11(18-5-4-12(13)21)7-24-15-19-9-3-2-8(23-14(16)17)6-10(9)20-15/h2-6,14H,7H2,1H3,(H,18,21)(H,19,20)
Formel vikt 353.0646