Läs mer
4’-O-Desmethyl 4’-O-Acetyl Diltiazem, TRC
Produktkod. 30354518
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354518 25 mg
30352246 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30354518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29070525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4'-O-Desmethyl 4'-O-Acetyl Diltiazem
CAS 85365-35-1
Molekylformel C23H26N2O5S
Kvantitet 25 mg
Synonym (2S,3S)-3-(Acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-acetyloxyphenyl)-1,5-benzothiazepin-4(5H)-one, (2S-cis)-3-(Acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-acetyloxyphenyl)-1,5-benzothiazepin-4(5H)-one
InChI formel InChI=1S/C23H26N2O5S/c1-15(26)29-18-11-9-17(10-12-18)22-21(30-16(2)27)23(28)25(14-13-24(3)4)19-7-5-6-8-20(19)31-22/h5-12,21-22H,13-14H2,1-4H3/t21-,22+/m1/s1
LEDER CN(C)CCN1C(=O)[C@H](OC(=O)C)[C@@H](Sc2ccccc12)c3ccc(OC(=O)C)cc3
Molekylvikt (g/mol) 442.53